Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr64f
- Entrez Gene ID
-
117477
- Gene Name
- Gustatory receptor 64f
- Gene Aliases
-
CG14988, CG32255, DmGr64f, Dmel64f, Dmel\CG32255, Dmel_CG32255, GR64f, Gr64, Gr64A3
- Chromosome Location
-
Chr.3L: 4035799 - 4037575 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052255
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168052.3 |
NP_728924.2 |
4 - 5 |
908 |
71 |
View Full Product Details