Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dyl
Entrez Gene ID
38531
Gene Name
dusky-like
Gene Aliases
CG15013, Dmel\CG15013, Dmel_CG15013, Dyl
Chromosome Location
Chr.3L: 4294004 - 4307718 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0066365
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139633.3 NP_647890.2 2 - 3 396 89
NM_168078.2 NP_728979.1 2 - 3 596 89
NM_168079.2 NP_728980.1 3 - 4 484 89
GenBank mRNA BT133367.1 - 2 - 3 308 89

View Full Product Details