Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NT1
Entrez Gene ID
38558
Gene Name
Neurotrophin 1
Gene Aliases
CG18318, CG32242, CG32244, CG42576, CT30097, DNT1, Dmel\CG42576, Dmel_CG42576, SPZ2, Spz-2, Spz2, dNT1, spz2
Chromosome Location
Chr.3L: 4501652 - 4508433 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261526
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169877.2 NP_001163348.1 4 - 5 1912 109
NM_001274488.1 NP_001261417.1 4 - 5 2024 109
NM_001274489.1 NP_001261418.1 4 - 5 1620 109
NM_168090.3 NP_729009.2 4 - 5 1912 109
GenBank mRNA FJ172423.1 - 4 - 5 1908 109

View Full Product Details