Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpr65Av
Entrez Gene ID
318014
Gene Name
Cuticular protein 65Av
Gene Aliases
BcDNA:RE31218, CG10461, CG32405, DmelCpr65Av, Dmel\CG32405, Dmel_CG32405
Chromosome Location
Chr.3L: 6139223 - 6139974 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052405
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_168158.2 NP_729146.1 1 - 2 259 64
GenBank mRNA BT001606.1 - 1 - 2 258 64
KX532120.1 - 1 - 2 146 64

View Full Product Details