Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or65a
- Entrez Gene ID
-
318011
- Gene Name
- Odorant receptor 65a
- Gene Aliases
-
65a, CG32401, DOR66, Dmel\CG32401, Dmel_CG32401, LU27.2, OR65a
- Chromosome Location
-
Chr.3L: 6320611 - 6322098 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041625
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168163.1 |
NP_729161.1 |
2 - 3 |
303 |
118 |
View Full Product Details