Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RhoGEF4
Entrez Gene ID
38806
Gene Name
Rho guanine nucleotide exchange factor 4
Gene Aliases
CG8606, DRhoGEF4, Dmel\CG8606, Dmel_CG8606, RhoGEF
Chromosome Location
Chr.3L: 7321099 - 7323411 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035761
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139844.3 NP_648101.1 2 - 3 796 58
NM_168199.2 NP_729232.1 1 - 2 597 58
GenBank mRNA AF255674.1 - 2 - 3 794 58
AY058496.1 - 2 - 3 765 58

View Full Product Details