Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- iav
- Entrez Gene ID
-
31621
- Gene Name
- inactive
- Gene Aliases
-
CG4536, CT14708, DmIav, Dmel\CG4536, Dmel_CG4536, IAV, Iav, TRPV, hypoB, osm-9
- Chromosome Location
-
Chr.X: 6813978 - 6818156 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086693
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132125.2 |
NP_572353.1 |
8 - 9 |
3362 |
61 |
View Full Product Details