Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bou
Entrez Gene ID
31644
Gene Name
boudin
Gene Aliases
BcDNA:RE28342, CG14430, Dmel\CG14430, Dmel_CG14430, JNXL4, jnL4, l(1)6Ea, l(1)jnL4
Chromosome Location
Chr.X: 6986486 - 6987188 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261284
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132145.4 NP_572373.1 1 - 1 553 73
GenBank mRNA AY071242.1 - 1 - 1 548 73

View Full Product Details