Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dok
- Entrez Gene ID
-
31667
- Gene Name
- Downstream of kinase
- Gene Aliases
-
CG2079, Ddok, Dmel\CG2079, Dmel_CG2079, dok, l(1)G0331
- Chromosome Location
-
Chr.X: 7324214 - 7328806 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0029944
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132163.4 |
NP_572391.1 |
1 - 2 |
459 |
80 |
GenBank mRNA |
AY061397.1 |
- |
1 - 2 |
455 |
80 |
View Full Product Details