Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sni
Entrez Gene ID
31761
Gene Name
sniffer
Gene Aliases
CG10964, Dmel\CG10964, Dmel_CG10964, SNI, Sni, anon-WO03040301.183
Chromosome Location
Chr.X: 8241501 - 8244016 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030026
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298083.1 NP_001285012.1 1 - 2 1072 63
NM_132238.3 NP_572466.1 1 - 2 416 63
GenBank mRNA AY118557.1 - 1 - 2 360 63

View Full Product Details