Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NaCP60E
Entrez Gene ID
37981
Gene Name
Na channel protein 60E
Gene Aliases
CG18506, CG34405, CG9071, CT24831, DIC60, DSC, DSC1, DSCI, Dmel\CG34405, Dmel_CG34405, Na-CP, dsc, dsc1, smi60E
Chromosome Location
Chr.2R: 24891614 - 24921677 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085434
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202078.3 NP_001189007.2 25 - 26 8376 84
NM_001259578.1 NP_001246507.1 22 - 23 7477 84
NM_001259580.2 NP_001246509.1 26 - 27 8391 84
NM_001274243.2 NP_001261172.1 25 - 26 8376 84
NM_001274244.2 NP_001261173.1 24 - 25 9581 84
NM_001274245.2 NP_001261174.1 21 - 22 9483 84
NM_001274246.2 NP_001261175.1 24 - 25 8483 84
NM_166696.5 NP_726495.3 25 - 26 8639 84

View Full Product Details