Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pain
Entrez Gene ID
37985
Gene Name
painless
Gene Aliases
CG15860, CT31987, Dmel\CG15860, Dmel_CG15860, EP2251, Pain
Chromosome Location
Chr.2R: 24921505 - 24928275 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0060296
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138135.2 NP_611979.1 2 - 3 254 82
GenBank mRNA AY268106.1 - 2 - 3 276 82
BT011483.1 - 2 - 3 251 82

View Full Product Details