Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
thoc7
Entrez Gene ID
38033
Gene Name
CG17143-PA
Gene Aliases
CG17143, Dmel\CG17143, Dmel_CG17143, THOC7, Thoc7
Chromosome Location
Chr.3L: 211636 - 212687 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035110
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138167.1 NP_612011.1 1 - 2 63 87
GenBank mRNA AJ620302.1 - 1 - 2 63 87

View Full Product Details