Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pyx
- Entrez Gene ID
-
38037
- Gene Name
- pyrexia
- Gene Aliases
-
CG17142, CT33412, Dmel\CG17142, Dmel_CG17142, Pyrexia, Pyx, dTRPA2, trpA2
- Chromosome Location
-
Chr.3L: 217932 - 223069 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035113
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138171.2 |
NP_612015.1 |
5 - 6 |
1549 |
57 |
GenBank mRNA |
BT010317.1 |
- |
5 - 6 |
1549 |
57 |
View Full Product Details