Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RhoGEF3
Entrez Gene ID
38050
Gene Name
Rho guanine nucleotide exchange factor 3
Gene Aliases
CG1225, CG13881, CG13882, CG13883, CG42377, CG42378, CG43976, Dmel\CG43976, Dmel_CG43976, DrhoGEF3, RhoGEF, rhoGEF3
Chromosome Location
Chr.3L: 277437 - 305292 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264707
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274273.1 NP_001261202.1 8 - 9 7654 82
NM_138180.4 NP_612024.4 8 - 9 7657 82

View Full Product Details