Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mthl10
Entrez Gene ID
38057
Gene Name
methuselah-like 10
Gene Aliases
CG17061, Dmel\CG17061, Dmel_CG17061, Mthl10, anon-WO0170980.166, anon-WO0170980.167, mth2, mthl-10
Chromosome Location
Chr.3L: 332801 - 342915 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035132
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259604.1 NP_001246533.1 3 - 4 1989 64
NM_001259605.2 NP_001246534.1 3 - 4 1989 64
NM_001274276.1 NP_001261205.1 3 - 4 1911 64
NM_001274277.1 NP_001261206.1 3 - 4 1481 64
NM_138186.3 NP_612030.2 3 - 4 1481 64

View Full Product Details