Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Trh
- Entrez Gene ID
-
38121
- Gene Name
- Tryptophan hydroxylase
- Gene Aliases
-
CG9122, CT9937, DTRH, DTRHn, Dmel\CG9122, Dmel_CG9122, TRH, TRHn, TrH, dTrh, trh
- Chromosome Location
-
Chr.3L: 1190384 - 1193173 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035187
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138236.2 |
NP_612080.1 |
1 - 2 |
596 |
63 |
GenBank mRNA |
BT001418.1 |
- |
1 - 2 |
572 |
63 |
View Full Product Details