Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
robl62A
Entrez Gene ID
38170
Gene Name
CG1014-PA
Gene Aliases
CG1014, Dmel\CG1014, Dmel_CG1014, rob162A
Chromosome Location
Chr.3L: 1477619 - 1478369 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028567
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138266.2 NP_612110.1 1 - 1 701 91

View Full Product Details