Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spz5
Entrez Gene ID
38350
Gene Name
spatzle 5
Gene Aliases
CG9972, CT28115, DNT2, Dmel\CG9972, Dmel_CG9972, NT2, Spz-5, Spz5
Chromosome Location
Chr.3L: 2883641 - 2892166 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035379
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139496.3 NP_647753.1 3 - 4 601 57
GenBank mRNA AY051735.1 - 3 - 4 583 57

View Full Product Details