Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- spz5
- Entrez Gene ID
-
38350
- Gene Name
- spatzle 5
- Gene Aliases
-
CG9972, CT28115, DNT2, Dmel\CG9972, Dmel_CG9972, NT2, Spz-5, Spz5
- Chromosome Location
-
Chr.3L: 2883641 - 2892166 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035379
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139496.3 |
NP_647753.1 |
5 - 6 |
1379 |
65 |
GenBank mRNA |
AY051735.1 |
- |
5 - 6 |
1361 |
65 |
View Full Product Details