Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cht7
Entrez Gene ID
38370
Gene Name
Chitinase 7
Gene Aliases
CG1869, DmCHT7, DmCht7, Dmel\CG1869, Dmel_CG1869
Chromosome Location
Chr.3L: 3090985 - 3106958 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035398
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139511.4 NP_647768.3 3 - 4 882 92
GenBank mRNA AY051988.1 - 3 - 4 855 92

View Full Product Details