Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sc2
- Entrez Gene ID
-
38457
- Gene Name
- CG10849-PA
- Gene Aliases
-
CG10849, Dmel\CG10849, Dmel_CG10849, SC2, Ter[CG10849], dSc2, l(3)05634, l(3)63Eb, l(3)SH4
- Chromosome Location
-
Chr.3L: 3903492 - 3905035 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035471
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139579.3 |
NP_647836.2 |
1 - 2 |
423 |
67 |
View Full Product Details