Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SMSr
- Entrez Gene ID
-
38823
- Gene Name
- CG32380-PA
- Gene Aliases
-
CG32380, CG8572, CG8576, Css3beta, Dmel\CG32380, Dmel_CG32380, dSMSr, dmSMSr
- Chromosome Location
-
Chr.3L: 7374050 - 7381474 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052380
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139857.3 |
NP_648114.1 |
2 - 3 |
1355 |
77 |
GenBank mRNA |
AY051605.1 |
- |
2 - 3 |
1398 |
77 |
View Full Product Details