Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tsp66A
- Entrez Gene ID
-
38836
- Gene Name
- Tetraspanin 66A
- Gene Aliases
-
CG16991, Dm.Tsp66A, Dmel\CG16991, Dmel_CG16991
- Chromosome Location
-
Chr.3L: 7465509 - 7468808 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035786
- Gene Symbol
- CG42660
- Entrez Gene ID
-
10178939
- Gene Name
- CG42660-PA
- Gene Aliases
-
Dmel\CG42660, Dmel_CG42660
- Chromosome Location
-
Chr.3L: 7465509 - 7468808 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261536
- Gene Symbol
- CG42661
- Entrez Gene ID
-
10178846
- Gene Name
- CG42661-PA
- Gene Aliases
-
Dmel\CG42661, Dmel_CG42661
- Chromosome Location
-
Chr.3L: 7465509 - 7468808 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261537
View Full Product Details