Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fry
Entrez Gene ID
39122
Gene Name
furry
Gene Aliases
CG14171, CG32045, CG6774, CG6780, Dmel\CG32045, Dmel_CG32045, Fry, RE-B, SS3-7, l(3)02240, yeti
Chromosome Location
Chr.3L: 9631834 - 9679103 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016081
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259770.1 NP_001246699.1 4 - 5 4019 75
NM_001259771.1 NP_001246700.1 4 - 5 3660 75
NM_001259772.1 NP_001246701.1 4 - 5 3660 75
NM_001274708.1 NP_001261637.1 4 - 5 4019 75
NM_001274709.1 NP_001261638.1 4 - 5 3660 75
NM_001274710.1 NP_001261639.1 4 - 5 3660 75
NM_168356.2 NP_729519.2 4 - 5 3660 75
NM_168357.3 NP_729520.1 4 - 5 3660 75

View Full Product Details