Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir67c
- Entrez Gene ID
-
317838
- Gene Name
- Ionotropic receptor 67c
- Gene Aliases
-
CG32058, DmelIR67c, Dmel\CG32058, Dmel_CG32058, IR67c
- Chromosome Location
-
Chr.3L: 10432384 - 10434105 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052058
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168407.1 |
NP_729609.1 |
1 - 1 |
1548 |
83 |
View Full Product Details