Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lola
Entrez Gene ID
44548
Gene Name
longitudinals lacking
Gene Aliases
BEST:LD03274, BTB-IV, BcDNA:LD17006, BcDNA:RH31485, BtbIV, CG12052, CG18376, CG18378, CG18379, CG18380, CG18381, CG30012, CG30013, CG30014, Dmel\CG12052, Dmel_CG12052, LD03274, Lola, NEST:bs06a08, bs06a08.y1, btb, eyeful, fus7, l(2)00642, l(2)s3697, sw59
Chromosome Location
Chr.2R: 10481894 - 10543291 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283521
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080027.6 NP_524766.2 5 - 6 1913 62
NM_176132.4 NP_788312.2 5 - 6 1974 62

View Full Product Details