Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp4e1
- Entrez Gene ID
-
44632
- Gene Name
- Cytochrome P450-4e1
- Gene Aliases
-
4e1, CG2062, CYP4E1, CYP4e1, Cyp4el, Dmel\CG2062, Dmel_CG2062, P-450, P450
- Chromosome Location
-
Chr.2R: 8447451 - 8449745 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015034
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080032.3 |
NP_524771.1 |
3 - 4 |
677 |
64 |
GenBank mRNA |
AY118793.1 |
- |
3 - 4 |
658 |
64 |
View Full Product Details