Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- kto
- Entrez Gene ID
-
44830
- Gene Name
- kohtalo
- Gene Aliases
-
Brm @ Kto, CG8491, Dmel\CG8491, Dmel_CG8491, KTO, Kto, Kto/Med12, MED12, Med12, SRB8, Srb8/KTO/TRAP230, Trap230, dTRAP230, dTrap230, l(3)76BDc, med12, med12/kto, psg14, siren1
- Chromosome Location
-
Chr.3L: 19836614 - 19844944 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001324
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080047.2 |
NP_524786.1 |
5 - 6 |
5738 |
65 |
GenBank mRNA |
AF324426.1 |
- |
5 - 6 |
5738 |
65 |
View Full Product Details