Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nkd
Entrez Gene ID
44843
Gene Name
naked cuticle
Gene Aliases
CG11614, CG18224, Dmel\CG11614, Dmel_CG11614, Nkd, naked
Chromosome Location
Chr.3L: 19003741 - 19045449 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002945
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275100.1 NP_001262029.1 2 - 3 1606 86
NM_001275101.1 NP_001262030.1 2 - 3 1606 86
NM_080049.3 NP_524788.2 2 - 3 1606 86
GenBank mRNA BT015258.1 - 2 - 3 1571 86

View Full Product Details