Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cht6
Entrez Gene ID
31935
Gene Name
Chitinase 6
Gene Aliases
CG15313, CG15314, CG2989, CG43374, CT5624, DmCht6, Dmel\CG43374, Dmel_CG43374
Chromosome Location
Chr.X: 10016845 - 10050803 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263132
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103464.3 NP_001096934.2 4 - 5 964 68
NM_001258669.2 NP_001245598.1 5 - 6 943 68
NM_001258670.2 NP_001245599.1 4 - 5 964 68
NM_001258671.2 NP_001245600.1 4 - 5 964 68
NM_001258672.2 NP_001245601.1 4 - 5 964 68
NM_001258673.2 NP_001245602.1 5 - 6 1020 68
NM_132370.4 NP_572598.3 4 - 5 964 68

View Full Product Details