Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psf3
Entrez Gene ID
31967
Gene Name
Partner of Sld Five 3
Gene Aliases
CG2222, Dmel\CG2222, Dmel_CG2222, PSF3
Chromosome Location
Chr.X: 10383353 - 10385861 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030196
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272477.1 NP_001259406.1 2 - 3 592 78
NM_001298154.1 NP_001285083.1 2 - 3 1304 78
NM_132396.2 NP_572624.1 2 - 3 592 78
GenBank mRNA AY089361.1 - 2 - 3 577 78

View Full Product Details