Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vago
Entrez Gene ID
32040
Gene Name
CG2081-PB
Gene Aliases
BcDNA:RH70969, CG2081, DIGF-2, Dmel\CG2081, Dmel_CG2081, VAGO, vago
Chromosome Location
Chr.X: 11089290 - 11090219 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030262
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298177.1 NP_001285106.1 1 - 2 163 78
NM_132452.3 NP_572680.1 1 - 2 163 78

View Full Product Details