Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FucT6
Entrez Gene ID
32122
Gene Name
alpha1,6-fucosyltransferase
Gene Aliases
6-FucT, BEST:CK00490, CG2448, CK00490, Dm alpha1, Dmel\CG2448, Dmel_CG2448, Fuc-TVIII, dalpha6Fut
Chromosome Location
Chr.X: 11706575 - 11709951 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030327
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132512.4 NP_572740.1 3 - 4 1722 80
GenBank mRNA AF441264.1 - 3 - 4 1218 80
AY051451.1 - 3 - 4 1673 80

View Full Product Details