Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP7
Entrez Gene ID
32142
Gene Name
ATP7-PB
Gene Aliases
Atp7, Atp7A, CG1886, DmATP7, Dmel\CG1886, Dmel_CG1886, atp7, dmATP7, lincRNA.S9122
Chromosome Location
Chr.X: 11855022 - 11864754 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030343
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272537.2 NP_001259466.1 2 - 3 2544 63
NM_132528.3 NP_572756.3 2 - 3 2544 63

View Full Product Details