Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sclp
Entrez Gene ID
32156
Gene Name
CG2471-PA
Gene Aliases
CG2471, CT8171, Dmel\CG2471, Dmel_CG2471, dSCLP
Chromosome Location
Chr.X: 11920004 - 11924662 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030357
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169256.1 NP_001162727.1 2 - 3 310 69
NM_001272546.1 NP_001259475.1 2 - 3 642 69
NM_132542.3 NP_572770.1 2 - 3 642 69
GenBank mRNA AY061562.1 - 2 - 3 642 69
BT072930.1 - 2 - 3 310 69
BT132925.1 - 2 - 3 642 69

View Full Product Details