Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec16
Entrez Gene ID
32209
Gene Name
Secretory 16
Gene Aliases
BcDNA:RE70141, CG18079, CG2551, CG32654, Dmel\CG32654, Dmel_CG32654, dSec16, dsec16, sec16, sec26
Chromosome Location
Chr.X: 12596047 - 12609910 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052654
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103493.4 NP_001096963.1 4 - 5 4354 90
NM_001103494.2 NP_001096964.1 4 - 5 4473 90
NM_001298228.1 NP_001285157.1 4 - 5 4473 90
NM_001298229.1 NP_001285158.1 5 - 6 4788 90
NM_001298230.1 NP_001285159.1 5 - 6 4788 90
NM_132584.3 NP_572812.3 5 - 6 4662 90

View Full Product Details