Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Brms1
Entrez Gene ID
32243
Gene Name
BRMS1 transcriptional repressor
Gene Aliases
BRMS1L, CG4400, Dmel\CG4400, Dmel_CG4400, dBrms1
Chromosome Location
Chr.X: 12907986 - 12909622 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030434
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132612.2 NP_572840.1 2 - 3 620 67
GenBank mRNA AY102677.1 - 2 - 3 597 67

View Full Product Details