Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REG
Entrez Gene ID
32274
Gene Name
Proteasome regulator gamma
Gene Aliases
CG1591, DmREGgamma, Dmel\CG1591, Dmel_CG1591, REGgamma, dREG, dREGgamma, dReg, dReggamma
Chromosome Location
Chr.X: 13212325 - 13214408 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029133
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272573.2 NP_001259502.1 2 - 3 574 93
NM_132638.4 NP_572866.1 2 - 3 547 93
GenBank mRNA AF215730.1 - 1 - 2 367 93
AY069688.1 - 2 - 3 532 93

View Full Product Details