Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lsd-2
- Entrez Gene ID
-
32437
- Gene Name
- Lipid storage droplet-2
- Gene Aliases
-
145098_at, CG9057, DmPLIN2, Dmel\CG9057, Dmel_CG9057, LSD, LSD-2, LSD2, LSP-2, Lsd2, PLIN2, Plin2, dmPLIN2, lsd-2, lsd2, plin2
- Chromosome Location
-
Chr.X: 15069127 - 15076220 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030608
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132768.3 |
NP_572996.1 |
3 - 4 |
958 |
77 |
GenBank mRNA |
AY060242.1 |
- |
3 - 4 |
941 |
77 |
View Full Product Details