Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab3-GEF
Entrez Gene ID
32442
Gene Name
Rab3 GDP-GTP exchange factor
Gene Aliases
BEST:GH21304, CG5627, Dmel\CG5627, Dmel_CG5627, anon-EST:Posey246, rab3-GEF, rab3-gef
Chromosome Location
Chr.X: 15089682 - 15113762 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030613
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103513.3 NP_001096983.2 11 - 12 4058 76
NM_001258746.2 NP_001245675.1 12 - 13 4076 76
NM_001258747.1 NP_001245676.1 11 - 12 4034 76
NM_001272630.1 NP_001259559.1 11 - 12 4034 76
NM_001272631.1 NP_001259560.1 12 - 13 4067 76
NM_001316635.1 NP_001303564.1 12 - 13 4067 76
NM_001347809.1 NP_001334679.1 13 - 14 4118 76
NM_001347810.1 NP_001334680.1 13 - 14 4118 76
NM_001347811.1 NP_001334681.1 13 - 14 4118 76
NM_132773.3 NP_573001.2 11 - 12 4034 76

View Full Product Details