Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sing
- Entrez Gene ID
-
32644
- Gene Name
- singles bar
- Gene Aliases
-
CG13011, Dmel\CG13011, Dmel_CG13011, Sing, singles-bar
- Chromosome Location
-
Chr.X: 16679297 - 16681784 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261245
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132918.2 |
NP_573146.1 |
2 - 3 |
477 |
68 |
View Full Product Details