Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk23
Entrez Gene ID
32731
Gene Name
pickpocket 23
Gene Aliases
CG8527, Dmel\CG8527, Dmel_CG8527, PPK-23, PPK23, Ppk23
Chromosome Location
Chr.X: 17566552 - 17569228 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030844
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272717.1 NP_001259646.1 4 - 5 1135 73
NM_132992.3 NP_573220.2 4 - 5 1135 73

View Full Product Details