Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HDAC1
- Entrez Gene ID
-
38565
- Gene Name
- Histone deacetylase 1
- Gene Aliases
-
CG7471, DHDAC1, DRpd3, DmHDAC1, DmRpd3, Dmel\CG7471, Dmel_CG7471, E(var)3-64BC, HDAC, HDAC-1, Hdac, Hdac1, Hdac1/Rpd3, RPD3, Rpd3, Rpd3/HDAC, Su(var)3-26, Su(var)3-64B, Su(var)326, Su(var)328, dHDAC-1, dHDAC1, dRPD3, dRpd3, dmHDA401, drpd3, hdac1, l(3)04556, l(3)64Cc, rpd3, rpd[3]
- Chromosome Location
-
Chr.3L: 4626734 - 4629467 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015805
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139661.4 |
NP_647918.2 |
3 - 4 |
1611 |
58 |
View Full Product Details