Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spo
Entrez Gene ID
38631
Gene Name
spook
Gene Aliases
307a1, CG10594, CYP307A1, CYP307A1-Dm, Cyp307a1, Dmel\CG10594, Dmel_CG10594, Dmspo, Spo
Chromosome Location
Chr.3L: 5590083 - 5592571 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003486
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300014.1 NP_001286943.1 1 - 1 1939 69
NM_139718.3 NP_647975.2 1 - 1 1637 69
GenBank mRNA AF484415.1 - 1 - 1 1627 69
BT133345.1 - 1 - 1 1939 69

View Full Product Details