Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jon65Ai
Entrez Gene ID
38685
Gene Name
Jonah 65Ai
Gene Aliases
CG10475, CT29408, Dmel\CG10475, Dmel_CG10475, Jon65, Jon65A, SP145
Chromosome Location
Chr.3L: 6053493 - 6054443 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035667
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300022.1 NP_001286951.1 1 - 1 759 64
NM_139758.2 NP_648015.1 1 - 1 759 64
GenBank mRNA AY118988.1 - 1 - 1 742 64

View Full Product Details