Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eIF4E4
- Entrez Gene ID
-
38743
- Gene Name
- eukaryotic translation initiation factor 4E4
- Gene Aliases
-
BEST:GH01027, CG10124, CT28485, Dmel\CG10124, Dmel_CG10124, NEST:bs04e06, eIF4E, eIF4E-4
- Chromosome Location
-
Chr.3L: 6664224 - 6665641 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035709
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139795.3 |
NP_648052.1 |
1 - 2 |
470 |
74 |
GenBank mRNA |
BT021432.1 |
- |
1 - 2 |
462 |
74 |
View Full Product Details