Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhea
Entrez Gene ID
38978
Gene Name
CG6831-PB
Gene Aliases
CG6831, CT21159, Dmel\CG6831, Dmel_CG6831, Rhea, Talin, Tln, talin, tendrils
Chromosome Location
Chr.3L: 8545001 - 8574409 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260442
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259745.2 NP_001246674.1 15 - 16 8123 74
NM_001274657.1 NP_001261586.1 14 - 15 8476 74
NM_001274658.1 NP_001261587.1 15 - 16 8060 74
NM_001274660.1 NP_001261589.1 16 - 17 8261 74
NM_139981.4 NP_648238.1 14 - 15 7948 74
GenBank mRNA AF299248.1 - 13 - 14 7895 74

View Full Product Details