Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsf
Entrez Gene ID
37068
Gene Name
Heat shock factor
Gene Aliases
CG5748, D-HSF, Dm-Hsf, DmHSF, DmHSFa, Dmel\CG5748, Dmel_CG5748, HSF, HSF1, HSFa, Hsf1, dHSF, dHSFa, heat shock factor, hsf, l(2)03091
Chromosome Location
Chr.2R: 18130303 - 18134634 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001222
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043090.3 NP_001036555.1 7 - 8 2299 102
NM_001043091.3 NP_001036556.1 8 - 9 2240 102
NM_001043092.3 NP_001036557.1 6 - 7 2245 102
NM_057227.5 NP_476575.1 7 - 8 2173 102
GenBank mRNA AB211168.1 - 5 - 6 2002 102
AB211169.1 - 7 - 8 1984 102
AB211170.1 - 6 - 7 2056 102
M60070.1 - 7 - 8 2158 102

View Full Product Details