Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Khc
- Entrez Gene ID
-
36810
- Gene Name
- Kinesin heavy chain
- Gene Aliases
-
CG7765, DK, DKH, Dm KHC, DmK, DmKHC, Dmel\CG7765, Dmel_CG7765, Dmkin, KHC, KIF 5A, KIF5, KIF5B, KIN, Kif5, Kin, Kin-1, Kinesin-1, khc, kin, kinesin, kinesin-1, l(2)W12, l(2)k13219, l(2)k13314, l(2R)W12, pgs
- Chromosome Location
-
Chr.2R: 16266960 - 16271971 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001308
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057242.5 |
NP_476590.1 |
1 - 2 |
629 |
69 |
GenBank mRNA |
AY094959.1 |
- |
1 - 2 |
636 |
69 |
View Full Product Details